Gene Transcripts

Gene Symbol
MTMR6
Entrez Gene ID
9107
Gene Name
myotubularin related protein 6
Gene Aliases
Chromosome Location
Chr. 13: 25246201 - 25287566 on Build GRCh38
UniGene ID
Hs.643702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004685.3 NP_004676.3 5 836
XM_011535306.1 XP_011533608.1 5 782
XM_011535307.1 XP_011533609.1 5 782
XM_017020845.1 XP_016876334.1 5 782
GenBank mRNA AF072928.1 - 1 50
AF406619.1 - 5 517
AK314587.1 - 5 701
AL832017.2 - 5 759
BC040012.1 - 5 708 5495564

View Full Product Details