Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002031.2 | NP_002022.1 | 1 | 521 | |
XM_005266880.4 | XP_005266937.1 | 2 | 593 | ||
XM_005266881.2 | XP_005266938.1 | 2 | 510 | ||
XM_005266882.4 | XP_005266939.1 | 3 | 822 | ||
XM_011535653.2 | XP_011533955.1 | 3 | 825 | ||
XM_011535654.2 | XP_011533956.1 | 2 | 607 | ||
XM_011535655.2 | XP_011533957.1 | 2 | 604 | ||
XM_017010645.1 | XP_016866134.1 | 2 | 711 | ||
GenBank mRNA | AK313314.1 | - | 1 | 601 | |
BC012916.1 | - | 1 | 366 | 4515877 | |
U00803.1 | - | 1 | 521 | ||
U22322.1 | - | 1 | 483 |