Gene Transcripts

Gene Symbol
PDP1
Entrez Gene ID
54704
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 1
Gene Aliases
PDH, PDP, PDPC, PPM2A, PPM2C
Chromosome Location
Chr. 8: 93916855 - 93926068 on Build GRCh38
UniGene ID
Hs.22265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161779.1 NP_001155251.1 3 991
NM_001161780.1 NP_001155252.1 3 865
NM_001161781.1 NP_001155253.1 2 852
NM_018444.3 NP_060914.2 2 909
XM_011517135.2 XP_011515437.1 3 762
XM_011517136.1 XP_011515438.1 3 869
XM_017013587.1 XP_016869076.1 2 924
XM_017013588.1 XP_016869077.1 3 832
GenBank mRNA AF155661.1 - 3 879
AK057079.1 - 2 821
AK126862.1 - 1 1286
AK302651.1 - 2 795
BC047619.1 - 2 830 4824168
BC064978.1 - 3 849
BC098343.1 - 1 663 40010016

View Full Product Details