Gene Transcripts

Gene Symbol
TPRX1
Entrez Gene ID
284355
Gene Name
tetrapeptide repeat homeobox 1
Gene Aliases
TPRX
Chromosome Location
Chr. 19: 47801243 - 47803604 on Build GRCh38
UniGene ID
Hs.629812
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198479.2 NP_940881.2 2 1728
GenBank mRNA AK097640.1 - 2 1728

View Full Product Details