Gene Transcripts

Gene Symbol
MAZ
Entrez Gene ID
4150
Gene Name
MYC associated zinc finger protein
Gene Aliases
PUR1, Pur-1, SAF-1, SAF-2, SAF-3, ZF87, ZNF801, Zif87
Chromosome Location
Chr. 16: 29806096 - 29811183 on Build GRCh38
UniGene ID
Hs.23650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042539.2 NP_001036004.1 3 1226
NM_001276275.1 NP_001263204.1 4 1041
NM_002383.3 NP_002374.2 3 1226
NR_074080.1 2 375
XM_006721047.3 XP_006721110.1 4 1013
XM_006721048.3 XP_006721111.1 3 1386
GenBank mRNA AB209061.1 - 1 758
AF489858.1 - 3 1173
BC041629.1 - 2 376 4309842
BI198991.1 - 2 260
D85131.1 - 4 1227
EF059746.1 - 3 1061
FJ532357.1 - 4 1031
L01420.1 - 2 473
M93339.1 - 3 1112
M94046.1 - 3 1083
U33819.1 - 3 1241

View Full Product Details