Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005253.3 | NP_005244.1 | 4 | 2043 | |
XM_006711976.2 | XP_006712039.1 | 4 | 2247 | ||
XM_006711977.3 | XP_006712040.1 | 4 | 3100 | ||
XM_017003737.1 | XP_016859226.1 | 4 | 2756 | ||
GenBank mRNA | AK055579.1 | - | 4 | 2097 | |
BC022791.1 | - | 4 | 1467 | 4637727 | |
BX647822.1 | - | 4 | 1219 |