Gene Transcripts

Gene Symbol
ZNF16
Entrez Gene ID
7564
Gene Name
zinc finger protein 16
Gene Aliases
HZF1, KOX9
Chromosome Location
Chr. 8: 144930358 - 144950888 on Build GRCh38
UniGene ID
Hs.493225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029976.2 NP_001025147.2 3 364
NM_006958.2 NP_008889.2 2 278
XM_005272341.2 XP_005272398.1 2 255
XM_005272342.2 XP_005272399.1 2 265
XM_011517298.1 XP_011515600.1 2 380
GenBank mRNA AF244088.1 - 1 186
AK096815.1 - 2 265
AK127625.1 - 2 278
AK226016.1 - 2 248
AL834451.1 - 1 195
BC010996.2 - 2 212 3949487
DQ117529.1 - 3 364

View Full Product Details