Gene Transcripts

Gene Symbol
ZNF22
Entrez Gene ID
7570
Gene Name
zinc finger protein 22
Gene Aliases
HKR-T1, KOX15, ZNF422, Zfp422
Chromosome Location
Chr. 10: 45000825 - 45005329 on Build GRCh38
UniGene ID
Hs.462693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006963.4 NP_008894.2 2 1484
GenBank mRNA BC010642.1 - 2 1379
BC041139.1 - 2 1839 5751460
BC053687.1 - 2 1383 5214077

View Full Product Details