Gene Transcripts

Gene Symbol
BAZ1A
Entrez Gene ID
11177
Gene Name
bromodomain adjacent to zinc finger domain 1A
Gene Aliases
ACF1, WALp1, WCRF180, hACF1
Chromosome Location
Chr. 14: 34752731 - 34875647 on Build GRCh38
UniGene ID
Hs.509140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013448.2 NP_038476.2 10 1768
NM_182648.1 NP_872589.1 10 1768
XM_011536374.1 XP_011534676.1 9 1067
GenBank mRNA AB032252.1 - 10 1765
AB209255.1 - 2 115
AF161435.1 - 10 1455
AF213467.1 - 10 1366
AF221130.1 - 9 1200

View Full Product Details