Gene Transcripts

Gene Symbol
AOC1
Entrez Gene ID
26
Gene Name
amine oxidase, copper containing 1
Gene Aliases
ABP, ABP1, DAO, DAO1, KAO
Chromosome Location
Chr. 7: 150824875 - 150861291 on Build GRCh38
UniGene ID
Hs.647097
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001091.3 NP_001082.2 2 309
NM_001272072.1 NP_001259001.1 2 309
XM_017011944.1 XP_016867433.1 4 517
XM_017011945.1 XP_016867434.1 4 547
XM_017011946.1 XP_016867435.1 2 473
XM_017011947.1 XP_016867436.1 2 287
GenBank mRNA AK296638.1 - 2 269
AK314806.1 - 2 267
BC014093.2 - 2 262 4547491
BX648159.1 - 4 4432
M55602.1 - 2 252
U11862.1 - 2 309
U11863.1 - 2 284

View Full Product Details