Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172632.1 | NP_001166103.1 | 3 | 308 | |
NM_001172633.1 | NP_001166104.1 | 3 | 308 | ||
NM_002543.3 | NP_002534.1 | 3 | 308 | ||
GenBank mRNA | AB010710.1 | - | 3 | 255 | |
AB102861.1 | - | 3 | 194 | ||
AF035776.1 | - | 3 | 240 | ||
AK292124.1 | - | 3 | 249 | ||
AK295409.1 | - | 3 | 249 | ||
AK316353.1 | - | 2 | 38 | ||
AL551100.3 | - | 3 | 232 | ||
AX429248.1 | - | 3 | 255 | ||
BC022295.1 | - | 3 | 249 | 4722086 | |
BP219342.1 | - | 3 | 308 |