Gene Transcripts
- Gene Symbol
- PDGFA
- Entrez Gene ID
- 5154
- Gene Name
- platelet derived growth factor subunit A
- Gene Aliases
- PDGF-A, PDGF1
- Chromosome Location
- Chr. 7: 497245 - 520668 on Build GRCh38
- UniGene ID
- Hs.535898
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002607.5 |
NP_002598.4
|
3, 4 |
1102 |
|
NM_033023.4 |
NP_148983.1
|
3, 4 |
1102 |
|
XM_011515415.1 |
XP_011513717.1
|
4, 5 |
594 |
|
XM_011515416.1 |
XP_011513718.1
|
4, 5 |
598 |
|
XM_011515417.1 |
XP_011513719.1
|
4, 5 |
594 |
|
XM_011515418.1 |
XP_011513720.1
|
4, 5 |
598 |
|
XM_011515419.2 |
XP_011513721.1
|
3, 4 |
302 |
|
XM_017012289.1 |
XP_016867778.1
|
3, 4 |
335 |
|
GenBank mRNA |
A09204.1 |
- |
3, 4 |
295 |
|
AB451309.1 |
- |
3, 4 |
259 |
|
AB451439.1 |
- |
3, 4 |
259 |
|
AU118732.1 |
- |
3, 4 |
430 |
|
BC109246.1 |
- |
1, 2 |
1122 |
|
BE336646.1 |
- |
2, 3 |
216 |
|
X03795.1 |
- |
3, 4 |
646 |
|
X06374.1 |
- |
3, 4 |
662 |
|
View Full Product Details