Gene Transcripts

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
Chromosome Location
Chr. 21: 36750888 - 36990254 on Build GRCh38
UniGene ID
Hs.371350
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000411.6 NP_000402.3 5 653
NM_001242784.1 NP_001229713.1 5 1587
NM_001242785.1 NP_001229714.1 5 746
XM_005260953.3 XP_005261010.1 4 615
XM_005260955.3 XP_005261012.1 4 486
XM_005260956.3 XP_005261013.1 4 662
XM_006723994.2 XP_006724057.1 4 600
XM_006723995.1 XP_006724058.1 5 778
XM_011529538.1 XP_011527840.1 5 633
XM_011529539.2 XP_011527841.1 5 638
XM_011529540.2 XP_011527842.1 4 615
XM_011529541.2 XP_011527843.1 4 489
XM_017028330.1 XP_016883819.1 4 436
GenBank mRNA AJ001864.1 - 4 676
AK307940.1 - 4 614
AK314189.1 - 4 520
BC060787.1 - 5 611 30343493
D23672.2 - 5 644
D87328.1 - 5 1404
X80160.1 - 4 504

View Full Product Details