Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003786.3 | NP_003777.2 | 16 | 2128 | |
XM_005257763.3 | XP_005257820.1 | 15 | 1950 | ||
XM_011525422.2 | XP_011523724.1 | 15 | 2055 | ||
XM_011525423.1 | XP_011523725.1 | 16 | 2247 | ||
XM_011525424.2 | XP_011523726.1 | 11 | 1478 | ||
XM_011525425.2 | XP_011523727.1 | 15 | 1955 | ||
XM_017025265.1 | XP_016880754.1 | 12 | 1991 | ||
XM_017025266.1 | XP_016880755.1 | 16 | 2247 | ||
GenBank mRNA | AB010887.1 | - | 16 | 2062 | |
AB208954.1 | - | 16 | 2068 | ||
AF009670.1 | - | 16 | 2121 | ||
AF083552.1 | - | 16 | 2065 | ||
AF085690.1 | - | 16 | 2084 | ||
AF085691.1 | - | 16 | 2084 | ||
AF085692.1 | - | 16 | 2139 | ||
AF104943.1 | - | 16 | 2052 | ||
BC137347.1 | - | 16 | 2079 | 9021345 | |
BC137348.1 | - | 16 | 2079 | 9021346 | |
Y17151.2 | - | 16 | 2090 |