Gene Transcripts

Gene Symbol
MYH9
Entrez Gene ID
4627
Gene Name
myosin, heavy chain 9, non-muscle
Gene Aliases
BDPLT6, DFNA17, EPSTS, FTNS, MHA, NMHC-II-A, NMMHC-IIA, NMMHCA
Chromosome Location
Chr. 22: 36281277 - 36388067 on Build GRCh38
UniGene ID
Hs.474751
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002473.5 NP_002464.1 3, 4 764
XM_011530197.2 XP_011528499.1 4, 5 619
XM_017028803.1 XP_016884292.1 3, 4 519
XM_017028804.1 XP_016884293.1 3, 4 519
XM_017028805.1 XP_016884294.1 3, 4 519
XM_017028806.1 XP_016884295.1 3, 4 519
GenBank mRNA AB191263.1 - 3, 4 664
AB290175.1 - 3, 4 557
AK131080.1 - 3, 4 630
AK291609.1 - 3, 4 662
AK304840.1 - 3, 4 658
AL832639.2 - 1, 2 429
BC049849.1 - 3, 4 633
BC090921.1 - 3, 4 570
CR456526.1 - 3, 4 565
M69180.1 - 2, 3 503
M81105.1 - 3, 4 623

View Full Product Details