Gene Transcripts

Gene Symbol
CCNE1
Entrez Gene ID
898
Gene Name
cyclin E1
Gene Aliases
CCNE, pCCNE1
Chromosome Location
Chr. 19: 29811994 - 29824317 on Build GRCh38
UniGene ID
Hs.244723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001238.3 NP_001229.1 5 470
NM_001322259.1 NP_001309188.1 5 470
NM_001322261.1 NP_001309190.1 5 470
NM_001322262.1 NP_001309191.1 4 423
XM_011527440.2 XP_011525742.1 5 356
GenBank mRNA AK291549.1 - 5 470
AU137789.1 - 5 470
BC035498.2 - 5 460 3841192
KF672847.1 - 3 299
KF672848.1 - 5 470
KR134287.1 - 5 470
M73812.1 - 3 299
M74093.1 - 3 267

View Full Product Details