Gene Transcripts

Gene Symbol
PIK3C2B
Entrez Gene ID
5287
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta
Gene Aliases
C2-PI3K
Chromosome Location
Chr. 1: 204422630 - 204494815 on Build GRCh38
UniGene ID
Hs.497487
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002646.3 NP_002637.3 3 619
XM_005245257.2 XP_005245314.1 2 684
XM_005245258.3 XP_005245315.1 2 684
XM_011509630.2 XP_011507932.1 4 1478
XM_011509631.2 XP_011507933.1 3 795
XM_011509633.2 XP_011507935.1 2 684
XM_017001473.1 XP_016856962.1 3 1614
XM_017001474.1 XP_016856963.1 3 817
GenBank mRNA AB210006.1 - 1 173
AK308153.1 - 3 697
BC132876.1 - 3 591 40146898
BC136645.1 - 3 591 9020635
BC144342.1 - 3 591 9052862
Y11312.1 - 3 655
Y13892.1 - 1 156

View Full Product Details