Gene Transcripts

Gene Symbol
PRM1
Entrez Gene ID
5619
Gene Name
protamine 1
Gene Aliases
CT94.1, P1
Chromosome Location
Chr. 16: 11280836 - 11281335 on Build GRCh38
UniGene ID
Hs.2909
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK311987.1 - 2 226
AY651260.1 - 2 214
BC003673.1 - 2 226 3935638
BT006746.1 - 2 136
CD360017.1 - 2 207
Y00443.1 - 2 236

View Full Product Details