Gene Transcripts

Gene Symbol
CHI3L2
Entrez Gene ID
1117
Gene Name
chitinase 3 like 2
Gene Aliases
CHIL2, YKL-39, YKL39
Chromosome Location
Chr. 1: 111227659 - 111243440 on Build GRCh38
UniGene ID
Hs.514840
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025197.1 NP_001020368.1 2 300
NM_001025199.1 NP_001020370.1 2 360
NM_004000.2 NP_003991.2 3 330
GenBank mRNA AK223316.1 - 2 240
AK298466.1 - 2 251
BC011460.1 - 3 268 4214121
BG698401.1 - 3 330
BM787868.1 - 2 350
BT006767.1 - 3 259
U49835.1 - 3 279
U58514.1 - 3 295
U58515.1 - 2 361

View Full Product Details