Gene Transcripts

Gene Symbol
SLC26A8
Entrez Gene ID
116369
Gene Name
solute carrier family 26 member 8
Gene Aliases
SPGF3, TAT1
Chromosome Location
Chr. 6: 35943514 - 36024855 on Build GRCh38
UniGene ID
Hs.435836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193476.1 NP_001180405.1 5 613
NM_052961.3 NP_443193.1 5 637
NM_138718.2 NP_619732.2 5 675
XM_011514294.2 XP_011512596.1 5 856
XM_017010235.1 XP_016865724.1 5 663
GenBank mRNA AF331522.1 - 5 581
AK122928.1 - 5 613
AK125189.1 - 5 857
BC025408.2 - 5 668 4823098
BG702985.1 - 5 607

View Full Product Details