Gene Transcripts

Gene Symbol
AQP7
Entrez Gene ID
364
Gene Name
aquaporin 7
Gene Aliases
AQP7L, AQPap, GLYCQTL
Chromosome Location
Chr. 9: 33383135 - 33402682 on Build GRCh38
UniGene ID
Hs.455323
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170.2 NP_001161.1 5 625
NM_001318156.1 NP_001305085.1 4 507
NM_001318157.1 NP_001305086.1 5 943
NM_001318158.1 NP_001305087.1 5 625
NR_134513.1 4 501
NR_134514.1 4 501
NR_134515.1 5 652
XM_005251453.3 XP_005251510.1 6 725
XM_006716765.3 XP_006716828.1 4 386
XM_011517866.2 XP_011516168.1 5 535
XM_011517867.2 XP_011516169.1 3 165
XM_017014700.1 XP_016870189.1 5 826
XM_017014701.1 XP_016870190.1 4 496
XM_017014702.1 XP_016870191.1 5 531
XM_017014703.1 XP_016870192.1 4 284
XM_017014704.1 XP_016870193.1 3 592
XM_017014705.1 XP_016870194.1 3 262
XM_017014706.1 XP_016870195.1 6 656
GenBank mRNA AB006190.1 - 5 462
AK294028.1 - 5 436
AK297860.1 - 5 530
AK297885.1 - 5 759
AK300716.1 - 2 403
AK301949.1 - 4 464
AK310456.1 - 3 364
AK315879.1 - 2 394
BC062701.1 - 4 334 30324263
BC119672.1 - 4 333
BC119673.1 - 4 333
DC306826.1 - 4 501

View Full Product Details