Gene Transcripts

Gene Symbol
RIT1
Entrez Gene ID
6016
Gene Name
Ras like without CAAX 1
Gene Aliases
NS8, RIBB, RIT, ROC1
Chromosome Location
Chr. 1: 155897808 - 155911402 on Build GRCh38
UniGene ID
Hs.491234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256820.1 NP_001243749.1 2 194
NM_001256821.1 NP_001243750.1 3 221
NM_006912.5 NP_008843.1 3 343
GenBank mRNA AF084462.1 - 3 254
AF493923.1 - 2 140
AK298768.1 - 2 194
AK314239.1 - 3 319
BC104186.2 - 3 214 40030939
BC104187.2 - 3 214 40030940
BI916790.1 - 3 204
BM546985.1 - 3 337
BQ918446.1 - 3 221
CR407639.1 - 2 140
U71203.1 - 3 275
U78165.1 - 3 275
Y07566.1 - 3 285

View Full Product Details