Gene Transcripts

Gene Symbol
RIT1
Entrez Gene ID
6016
Gene Name
Ras like without CAAX 1
Gene Aliases
NS8, RIBB, RIT, ROC1
Chromosome Location
Chr. 1: 155897808 - 155911402 on Build GRCh38
UniGene ID
Hs.491234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256820.1 NP_001243749.1 3, 4 291
NM_001256821.1 NP_001243750.1 4, 5 318
NM_006912.5 NP_008843.1 4, 5 440
GenBank mRNA AF084462.1 - 4, 5 351
AF493923.1 - 3, 4 237
AK298768.1 - 3, 4 291
AK314239.1 - 4, 5 416
BC104186.2 - 4, 5 311 40030939
BC104187.2 - 4, 5 311 40030940
BI916790.1 - 4, 5 301
BM546985.1 - 4, 5 434
BQ918446.1 - 4, 5 318
CR407639.1 - 3, 4 237
U71203.1 - 4, 5 372
U78165.1 - 4, 5 372
Y07566.1 - 4, 5 382

View Full Product Details