Gene Transcripts

Gene Symbol
SERPINB5
Entrez Gene ID
5268
Gene Name
serpin family B member 5
Gene Aliases
PI5, maspin
Chromosome Location
Chr. 18: 63476911 - 63505085 on Build GRCh38
UniGene ID
Hs.55279
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002639.4 NP_002630.2 7 981
XM_006722483.2 XP_006722546.1 4 489
GenBank mRNA AK301667.1 - 6 745
AK312765.1 - 7 959
BM955647.1 - 2 209
BX640597.1 - 2 2377
CV575332.1 - 4 468
U04313.1 - 7 914

View Full Product Details