Gene Transcripts

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location
Chr. 1: 16985958 - 17011972 on Build GRCh38
UniGene ID
Hs.128866
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001141973.2 NP_001135445.1 10 1072
NM_001141974.2 NP_001135446.1 10 1072
NM_022089.3 NP_071372.1 10 1087
XM_005245810.1 XP_005245867.1 10 1084
XM_005245811.1 XP_005245868.1 10 1072
XM_005245812.1 XP_005245869.1 10 1060
XM_005245815.1 XP_005245872.1 10 1087
XM_006710512.1 XP_006710575.1 10 1069
XM_006710513.1 XP_006710576.1 10 1045
XM_011541128.1 XP_011539430.1 10 1087
XM_011541129.1 XP_011539431.1 10 1087
XM_017000844.1 XP_016856333.1 10 1087
XM_017000845.1 XP_016856334.1 10 1069
XM_017000846.1 XP_016856335.1 10 1045
XM_017000847.1 XP_016856336.1 10 1057
XM_017000848.1 XP_016856337.1 10 1087
XM_017000849.1 XP_016856338.1 10 1072
XM_017000850.1 XP_016856339.1 10 1087
GenBank mRNA AK290210.1 - 10 1028
AL354615.1 - 10 887
AY461712.1 - 10 886
BC030267.1 - 10 1015 5240813
BC034575.1 - 7 578

View Full Product Details