Gene Transcripts

Gene Symbol
TAS1R3
Entrez Gene ID
83756
Gene Name
taste 1 receptor member 3
Gene Aliases
T1R3
Chromosome Location
Chr. 1: 1331324 - 1335320 on Build GRCh38
UniGene ID
Hs.74375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152228.2 NP_689414.1 3 1148
XM_017002435.1 XP_016857924.1 3 1170
XM_017002436.1 XP_016857925.1 3 1170

View Full Product Details