Gene Transcripts

Gene Symbol
WFS1
Entrez Gene ID
7466
Gene Name
wolframin ER transmembrane glycoprotein
Gene Aliases
CTRCT41, WFRS, WFS, WFSL
Chromosome Location
Chr. 4: 6260368 - 6303265 on Build GRCh38
UniGene ID
Hs.518602
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145853.1 NP_001139325.1 6, 7 881
NM_006005.3 NP_005996.2 6, 7 885
XM_017008586.1 XP_016864075.1 7, 8 864
GenBank mRNA AF084481.1 - 6, 7 885
AK312897.1 - 6, 7 814
BC030130.2 - 6, 7 815 4127654
KU178395.1 - 5, 6 715
Y18064.1 - 6, 7 873

View Full Product Details