Gene Transcripts

Gene Symbol
HMP19
Entrez Gene ID
51617
Gene Name
HMP19 protein
Gene Aliases
Chromosome Location
Chr. 5: 174045604 - 174109179 on Build GRCh38
UniGene ID
Hs.559412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015980.4 NP_057064.1 4, 5 579
GenBank mRNA AF113537.1 - 4, 5 448
AF123767.1 - 4, 5 448
AF133422.1 - 4, 5 432
AK294383.1 - 3, 4 362
AK295206.1 - 3, 4 262
AK297196.1 - 4, 5 385
AK312359.1 - 4, 5 466
AK315949.1 - 2, 3 194
AK315952.1 - 3, 4 276
BC002619.2 - 4, 5 462 3160345
BC018873.2 - 4, 5 455
CR457142.1 - 3, 4 317

View Full Product Details