Gene Transcripts

Gene Symbol
TYSND1
Entrez Gene ID
219743
Gene Name
trypsin domain containing 1
Gene Aliases
NET41
Chromosome Location
Chr. 10: 70137981 - 70146740 on Build GRCh38
UniGene ID
Hs.533655
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040273.2 NP_001035363.1 2 2019
NM_173555.3 NP_775826.2 4 2336
NR_073580.1 2 787
NR_073581.1 4 1224
NR_073582.1 4 1104
NR_073591.1 4 1015
NR_073592.1 2 907
NR_073593.1 4 1007
NR_073594.1 4 1127
NR_073595.1 4 1260
GenBank mRNA AK074968.1 - 4 1162
AK097697.1 - 3 1879
BC016840.1 - 4 1790
BC030242.1 - 2 1935 5176098
BC047424.1 - 2 787
BC111501.1 - 4 2275 5732356

View Full Product Details