Gene Transcripts
- Gene Symbol
- PI4KAP2
- Entrez Gene ID
- 375133
- Gene Name
- phosphatidylinositol 4-kinase alpha pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 22: 21472998 - 21517491 on Build GRCh38
- UniGene ID
- Hs.448225
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003700.1 |
|
17 |
2258 |
|
GenBank mRNA |
BC020225.1 |
- |
17 |
2257 |
|
- Gene Symbol
- PI4KA
- Entrez Gene ID
- 5297
- Gene Name
- phosphatidylinositol 4-kinase alpha
- Gene Aliases
- PI4K-ALPHA, PIK4CA, PMGYCHA, pi4K230
- Chromosome Location
- Chr. 22: 20707691 - 20858812 on Build GRCh38
- UniGene ID
- Hs.529438
- Species
- Homo sapiens
View Full Product Details