Gene Transcripts

Gene Symbol
ASAP3
Entrez Gene ID
55616
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 3
Gene Aliases
ACAP4, CENTB6, DDEFL1, UPLC1
Chromosome Location
Chr. 1: 23428563 - 23484631 on Build GRCh38
UniGene ID
Hs.437379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017707.3 NP_060177.2 4, 5 546
XM_011541755.1 XP_011540057.1 4, 5 480
XM_017001685.1 XP_016857174.1 4, 5 480
XM_017001686.1 XP_016857175.1 4, 5 480
XM_017001687.1 XP_016857176.1 4, 5 453
XM_017001688.1 XP_016857177.1 4, 5 442
XM_017001689.1 XP_016857178.1 4, 5 497
GenBank mRNA AB056749.1 - 4, 5 467
AK092291.1 - 4, 5 468
AK299359.1 - 4, 5 442
BC023519.2 - 5, 6 534
BC060786.1 - 4, 5 494 5298355

View Full Product Details