Gene Transcripts

Gene Symbol
ETNK1
Entrez Gene ID
55500
Gene Name
ethanolamine kinase 1
Gene Aliases
EKI, EKI 1, EKI1, Nbla10396
Chromosome Location
Chr. 12: 22625142 - 22690674 on Build GRCh38
UniGene ID
Hs.29464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018638.4 NP_061108.2 8 1726
GenBank mRNA AF207600.2 - 8 1792
AK074425.1 - 8 1671

View Full Product Details