Gene Transcripts
- Gene Symbol
- CDC42SE1
- Entrez Gene ID
- 56882
- Gene Name
- CDC42 small effector 1
- Gene Aliases
- SCIP1, SPEC1
- Chromosome Location
- Chr. 1: 151050971 - 151059649 on Build GRCh38
- UniGene ID
- Hs.22065
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF187845.2 |
- |
5 |
1162 |
|
AF286592.1 |
- |
4 |
1714 |
|
View Full Product Details