Gene Transcripts

Gene Symbol
CLMP
Entrez Gene ID
79827
Gene Name
CXADR like membrane protein
Gene Aliases
ACAM, ASAM, CSBM, CSBS
Chromosome Location
Chr. 11: 123072006 - 123195305 on Build GRCh38
UniGene ID
Hs.591949
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024769.3 NP_079045.1 4 809
XM_011542998.2 XP_011541300.1 4 777
GenBank mRNA AK026068.1 - 4 785
AY326422.1 - 4 633
AY358340.1 - 4 633
BC009371.2 - 4 455 4122648

View Full Product Details