Gene Transcripts

Gene Symbol
OVOS2
Entrez Gene ID
144203
Gene Name
ovostatin 2
Gene Aliases
Chromosome Location
Chr. 12: 31114623 - 31206154 on Build GRCh38
UniGene ID
Hs.524331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020288.1 XP_016875777.1 29 3916
GenBank mRNA AL831947.1 - 1967
BC039117.1 - 2 341
BC046190.1 - 2 191
CR749395.1 - 2 2155

Gene Symbol
LOC728715
Entrez Gene ID
728715
Gene Name
ovostatin homolog 2
Gene Aliases
Chromosome Location
Chr. 12: 9399917 - 9403421 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020286.1 XP_016875775.1 3 286

Gene Symbol
OVOS
Entrez Gene ID
408186
Gene Name
ovostatin
Gene Aliases
Chromosome Location
Chr. 12: 9517803 - 9573287 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020287.1 XP_016875776.1 28 3736
GenBank mRNA BX647938.1 - 9 6184

View Full Product Details