Gene Transcripts

Gene Symbol
TPRG1
Entrez Gene ID
285386
Gene Name
tumor protein p63 regulated 1
Gene Aliases
FAM79B
Chromosome Location
Chr. 3: 189000774 - 189325304 on Build GRCh38
UniGene ID
Hs.729544
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198485.3 NP_940887.1 2 303
XM_005247382.4 XP_005247439.1 2 3547
XM_005247388.3 XP_005247445.1 2 3559
XM_011512732.2 XP_011511034.2 2 2373
XM_011512734.2 XP_011511036.1 2 3553
XM_011512735.2 XP_011511037.1 2 3548
XM_011512736.2 XP_011511038.1 2 3559
XM_017006257.1 XP_016861746.1 9 1535
XM_017006258.1 XP_016861747.1 6 963
XM_017006259.1 XP_016861748.1 8 2146
XM_017006260.1 XP_016861749.1 8 1342
XM_017006261.1 XP_016861750.1 6 1151
XM_017006262.1 XP_016861751.1 1 147
GenBank mRNA AK123232.1 - 7 1362
AK125682.1 - 2 304
BC101550.1 - 2 204 8069056
BC101552.1 - 2 204 8069058

View Full Product Details