Gene Transcripts

Gene Symbol
CYTL1
Entrez Gene ID
54360
Gene Name
cytokine like 1
Gene Aliases
C17, C4orf4
Chromosome Location
Chr. 4: 5014586 - 5019470 on Build GRCh38
UniGene ID
Hs.13872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018659.2 NP_061129.1 4 611
XM_017008299.1 XP_016863788.1 5 726
GenBank mRNA AF193766.1 - 4 614
AF274956.1 - 4 471
AY359101.1 - 4 612
BC031391.1 - 4 587 5186334

View Full Product Details