Gene Transcripts

Gene Symbol
AGBL5
Entrez Gene ID
60509
Gene Name
ATP/GTP binding protein like 5
Gene Aliases
CCP5, RP75
Chromosome Location
Chr. 2: 27051423 - 27070622 on Build GRCh38
UniGene ID
Hs.138207
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035507.2 NP_001030584.1 10 1976
NM_021831.5 NP_068603.4 10 1976
NR_104246.1 10 1976
XM_005264477.2 XP_005264534.1 10 1976
XM_011533011.2 XP_011531313.1 10 1937
XM_011533012.2 XP_011531314.1 11 2142
XM_011533013.2 XP_011531315.1 11 1945
GenBank mRNA AK025492.1 - 8 1498
AK302091.1 - 10 1907
AL833844.1 - 10 1918
BC007415.2 - 10 1918 2967512
BC018562.1 - 1 51
BC018584.1 - 10 1921

View Full Product Details