Gene Transcripts

Gene Symbol
MFSD1
Entrez Gene ID
64747
Gene Name
major facilitator superfamily domain containing 1
Gene Aliases
SMAP4
Chromosome Location
Chr. 3: 158801870 - 158829719 on Build GRCh38
UniGene ID
Hs.58663
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167903.1 NP_001161375.1 6 747
NM_001289406.1 NP_001276335.1 6 751
NM_001289407.1 NP_001276336.1 7 833
NM_022736.2 NP_073573.2 7 864
NR_110328.1 6 751
XM_006713730.2 XP_006713793.1 6 682
GenBank mRNA AB014732.1 - 7 696
AF351617.2 - 7 560
AK024215.1 - 7 667
AK294628.1 - 6 624
AK297034.1 - 455
AK297593.1 - 7 864
AK300497.1 - 6 550
AK301680.1 - 7 603
BC030542.1 - 6 620 4915325
BC042197.1 - 7 676 5737259

View Full Product Details