Gene Transcripts

Gene Symbol
HGH1
Entrez Gene ID
51236
Gene Name
HGH1 homolog
Gene Aliases
BRP16, BRP16L, C8orf30A, C8orf30B, FAM203A, FAM203B
Chromosome Location
Chr. 8: 144137721 - 144140843 on Build GRCh38
UniGene ID
Hs.300224
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016458.3 NP_057542.2 6 1372
GenBank mRNA AF151022.1 - 3 764
AK098226.1 - 7 1066
BC003035.1 - 6 1372 2822563
BC009915.2 - 6 1372 2822563
BC015471.1 - 7 903

View Full Product Details