Gene Transcripts

Gene Symbol
RIBC1
Entrez Gene ID
158787
Gene Name
RIB43A domain with coiled-coils 1
Gene Aliases
2610028I09Rik
Chromosome Location
Chr. X: 53422782 - 53431120 on Build GRCh38
UniGene ID
Hs.351743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031745.4 NP_001026915.1 3 321
NM_001267053.3 NP_001253982.1 3 321
NM_144968.3 NP_659405.1 3 321
XM_005261988.3 XP_005262045.1 3 529
XM_005261990.3 XP_005262047.1 3 395
GenBank mRNA AK057345.1 - 3 287
AK290346.1 - 3 271
AK292871.1 - 3 276
AK304179.1 - 3 321
BC036767.1 - 3 236 5744127

View Full Product Details