Gene Transcripts

Gene Symbol
DCLK2
Entrez Gene ID
166614
Gene Name
doublecortin like kinase 2
Gene Aliases
CL2, CLICK-II, CLICK2, CLIK2, DCAMKL2, DCDC3, DCDC3B, DCK2
Chromosome Location
Chr. 4: 150078274 - 150258563 on Build GRCh38
UniGene ID
Hs.591683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040260.3 NP_001035350.2 2 1462
NM_001040261.4 NP_001035351.4 2 1462
NR_036614.1 2 1462
XM_005262785.2 XP_005262842.1 2 1291
XM_005262786.2 XP_005262843.1 2 1291
XM_017007832.1 XP_016863321.1 2 1291
XM_017007833.1 XP_016863322.1 2 1291
GenBank mRNA AB209181.1 - 2 1462
AL834498.1 - 2 509
BC032726.1 - 2 755

View Full Product Details