Gene Transcripts

Gene Symbol
NKAPP1
Entrez Gene ID
158801
Gene Name
NFKB activating protein pseudogene 1
Gene Aliases
CXorf42
Chromosome Location
Chr. X: 120236452 - 120245267 on Build GRCh38
UniGene ID
Hs.442518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027131.1 3 1173
GenBank mRNA AK093895.1 - 3 1139
BC022359.1 - 3 1113
BC113730.1 - 3 1112
BC113732.1 - 3 1112

View Full Product Details