Gene Transcripts

Gene Symbol
MACC1
Entrez Gene ID
346389
Gene Name
metastasis associated in colon cancer 1
Gene Aliases
7A5, SH3BP4L
Chromosome Location
Chr. 7: 20134655 - 20217390 on Build GRCh38
UniGene ID
Hs.598388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182762.3 NP_877439.3 5 478
GenBank mRNA AJ313524.1 - 2 169
AK131400.1 - 5 478
BC137090.1 - 3 199 9021087

View Full Product Details