Gene Transcripts
- Gene Symbol
- SLC25A25
- Entrez Gene ID
- 114789
- Gene Name
- solute carrier family 25 member 25
- Gene Aliases
- MCSC, PCSCL, SCAMC-2
- Chromosome Location
- Chr. 9: 128068200 - 128109258 on Build GRCh38
- UniGene ID
- Hs.5476
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001006641.3 |
NP_001006642.1
|
3, 4 |
604 |
|
NM_001006642.3 |
NP_001006643.1
|
3, 4 |
849 |
|
NM_052901.4 |
NP_443133.2
|
3, 4 |
467 |
|
NR_049766.2 |
|
3, 4 |
510 |
|
XM_005251689.4 |
XP_005251746.1
|
3, 4 |
863 |
|
GenBank mRNA |
AJ619963.1 |
- |
3, 4 |
768 |
|
AJ619989.1 |
- |
3, 4 |
392 |
|
AJ619990.1 |
- |
3, 4 |
487 |
|
AJ619991.1 |
- |
3, 4 |
449 |
|
AJ619992.1 |
- |
3, 4 |
435 |
|
AK290705.1 |
- |
3, 4 |
440 |
|
AK290991.1 |
- |
3, 4 |
604 |
|
AL559911.3 |
- |
3, 4 |
612 |
|
AY358515.1 |
- |
3, 4 |
442 |
|
BC089448.1 |
- |
3, 4 |
458 |
30558551 |
BC103930.1 |
- |
3, 4 |
409 |
40008034 |
BC103931.1 |
- |
3, 4 |
409 |
40008035 |
BC103932.1 |
- |
3, 4 |
409 |
40008036 |
BC103933.1 |
- |
3, 4 |
409 |
40008037 |
BQ637100.1 |
- |
3, 4 |
467 |
|
View Full Product Details