Gene Transcripts
- Gene Symbol
- SPTAN1
- Entrez Gene ID
- 6709
- Gene Name
- spectrin alpha, non-erythrocytic 1
- Gene Aliases
- EIEE5, NEAS, SPTA2
- Chromosome Location
- Chr. 9: 128552558 - 128633665 on Build GRCh38
- UniGene ID
- Hs.372331
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130438.2 |
NP_001123910.1
|
12, 13 |
1710 |
|
NM_001195532.1 |
NP_001182461.1
|
12, 13 |
1710 |
|
NM_003127.3 |
NP_003118.2
|
12, 13 |
1710 |
|
XM_006717245.1 |
XP_006717308.1
|
12, 13 |
1617 |
|
XM_006717246.1 |
XP_006717309.1
|
12, 13 |
1617 |
|
XM_006717247.2 |
XP_006717310.1
|
12, 13 |
1617 |
|
XM_006717248.2 |
XP_006717311.1
|
12, 13 |
1616 |
|
XM_006717249.1 |
XP_006717312.1
|
12, 13 |
1617 |
|
XM_006717250.1 |
XP_006717313.1
|
12, 13 |
1617 |
|
XM_006717251.2 |
XP_006717314.1
|
12, 13 |
1704 |
|
XM_006717252.2 |
XP_006717315.1
|
12, 13 |
1617 |
|
XM_006717253.1 |
XP_006717316.1
|
12, 13 |
1617 |
|
XM_006717254.2 |
XP_006717317.1
|
12, 13 |
1704 |
|
XM_017015059.1 |
XP_016870548.1
|
12, 13 |
1695 |
|
XM_017015060.1 |
XP_016870549.1
|
12, 13 |
1617 |
|
XM_017015061.1 |
XP_016870550.1
|
12, 13 |
1675 |
|
GenBank mRNA |
AB191262.1 |
- |
12, 13 |
1675 |
|
AB209860.1 |
- |
12, 13 |
1672 |
|
AK294753.1 |
- |
13, 14 |
1734 |
|
AK300386.1 |
- |
12, 13 |
1678 |
|
AK307676.1 |
- |
12, 13 |
1678 |
|
BC053521.1 |
- |
12, 13 |
1659 |
5529391 |
J05243.1 |
- |
12, 13 |
1670 |
|
U83867.1 |
- |
12, 13 |
1637 |
|
View Full Product Details