Gene Transcripts

Gene Symbol
KLHDC3
Entrez Gene ID
116138
Gene Name
kelch domain containing 3
Gene Aliases
PEAS, dJ20C7.3
Chromosome Location
Chr. 6: 43014103 - 43021298 on Build GRCh38
UniGene ID
Hs.412468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057161.3 NP_476502.1 11 1935
NR_040101.1 10 1819
GenBank mRNA AB055925.1 - 11 1799
AK097719.1 - 9 1661
BC000295.2 - 11 1755 2819570
BC001789.1 - 11 1770 3355470
BC001793.2 - 11 1733 3352397
BC007296.1 - 11 1785 3349731
BC009460.1 - 12 1417 3537223
BC021546.1 - 11 1772 3351635
BC041793.1 - 10 1673 5288540
BC045612.1 - 10 1673 5313255

View Full Product Details