Gene Transcripts

Gene Symbol
FAM3D
Entrez Gene ID
131177
Gene Name
family with sequence similarity 3 member D
Gene Aliases
EF7, OIT1
Chromosome Location
Chr. 3: 58633943 - 58666843 on Build GRCh38
UniGene ID
Hs.61265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138805.2 NP_620160.1 5 492
XM_005264862.4 XP_005264919.1 5 465
XM_005264863.4 XP_005264920.1 5 498
XM_005264864.4 XP_005264921.1 5 470
XM_006712965.3 XP_006713028.1 5 262
XM_006712967.3 XP_006713030.1 4 389
XM_011533348.2 XP_011531650.1 5 519
XM_011533349.2 XP_011531651.1 5 448
XM_011533350.2 XP_011531652.1 5 516
XM_011533351.2 XP_011531653.1 5 538
XM_011533352.2 XP_011531654.1 4 392
XM_011533353.2 XP_011531655.1 4 322
XM_011533354.2 XP_011531656.1 4 319
GenBank mRNA AB105189.1 - 5 495
AF494381.1 - 3 195
AK127546.1 - 5 487
AY358677.1 - 5 506
BC015359.1 - 5 401 3852062

View Full Product Details