Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286574.1 | NP_001273503.1 | 3, 4 | 529 | |
NM_001286576.1 | NP_001273505.1 | 3, 4 | 529 | ||
NM_145028.4 | NP_659465.2 | 3, 4 | 610 | ||
XM_011514381.2 | XP_011512683.1 | 3, 4 | 478 | ||
XM_011514382.2 | XP_011512684.1 | 3, 4 | 609 | ||
XM_017010435.1 | XP_016865924.1 | 4, 5 | 2084 | ||
GenBank mRNA | AK058119.1 | - | 3, 4 | 479 | |
BC033033.2 | - | 3, 4 | 560 | 5268250 | |
CR735810.1 | - | 3, 4 | 423 |