Gene Transcripts

Gene Symbol
SYNE3
Entrez Gene ID
161176
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, NET53, Nesp3
Chromosome Location
Chr. 14: 95416083 - 95516693 on Build GRCh38
UniGene ID
Hs.354645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152592.3 NP_689805.3 15, 16 2705
XM_005267377.3 XP_005267434.1 16, 17 2802
XM_006720063.3 XP_006720126.1 16, 17 2902
XM_011536515.2 XP_011534817.1 16, 17 2837
XM_017021056.1 XP_016876545.1 15, 16 2689
GenBank mRNA AK098471.1 - 10, 11 1468
AK131436.1 - 16, 17 2555
BC146604.1 - 16, 17 2702 40123293

View Full Product Details